siRNA Details
| Gene Symbol: Appbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025825.3 | NP_080101.1 | 5, 6 | 978 | ||
| GenBank mRNA | AK004888.1 | - | 5, 6 | 989 | ||
| AK005440.1 | - | 5, 6 | 684 | |||
| AK122222.1 | - | 585 | ||||
| AK133853.1 | - | 5, 6 | 970 | |||
| AK207679.1 | - | 1, 2 | 177 | |||
| AK207768.1 | - | 2, 3 | 287 | |||
| AK217461.1 | - | 2, 3 | 251 | |||
| BC018442.1 | - | 5, 6 | 956 | 3157290 | ||
| BC019152.1 | - | 5, 6 | 979 | 5065139 | ||

Appbp2
Genomic Map