siRNA Details
| Gene Symbol: Lman2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025828.3 | NP_080104.2 | 8 | 1835 | ||
| GenBank mRNA | AK004952.1 | - | 8 | 1666 | ||
| AK014384.1 | - | 8 | 1657 | |||
| AK083442.1 | - | 8 | 1668 | |||
| AK129028.1 | - | 8 | 1662 | |||
| AK169401.1 | - | 8 | 1663 | |||
| BC055327.1 | - | 8 | 1835 | 5702182 | ||

Lman2
Genomic Map