siRNA Details
| Gene Symbol: Pxdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025831.3 | NP_080107.3 | 2, 3 | 589 | ||
| GenBank mRNA | BC018520.1 | - | 2, 3 | 277 | ||
| BC037079.1 | - | 2, 3 | 588 | 5051974 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pxdc1| Gene Symbol: Pxdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025831.3 | NP_080107.3 | 2, 3 | 589 | ||
| GenBank mRNA | BC018520.1 | - | 2, 3 | 277 | ||
| BC037079.1 | - | 2, 3 | 588 | 5051974 | ||