siRNA Details
| Gene Symbol: Trim59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025863.3 | NP_080139.3 | 3 | 2595 | ||
| GenBank mRNA | AK009637.1 | - | 1 | 1668 | ||
| BC006700.1 | - | 3 | 2507 | 3501285 | ||
| BC025430.1 | - | 3 | 2521 | 4017983 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim59| Gene Symbol: Trim59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025863.3 | NP_080139.3 | 3 | 2595 | ||
| GenBank mRNA | AK009637.1 | - | 1 | 1668 | ||
| BC006700.1 | - | 3 | 2507 | 3501285 | ||
| BC025430.1 | - | 3 | 2521 | 4017983 | ||