siRNA Details
| Gene Symbol: Tmem206 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025864.3 | NP_080140.1 | 2 | 183 | ||
| GenBank mRNA | AK009526.1 | - | 2 | 184 | ||
| AK012107.1 | - | 2 | 201 | |||
| AK031270.1 | - | 2 | 217 | |||
| AK044482.1 | - | 2 | 184 | |||
| AK049641.1 | - | 2 | 220 | |||

Tmem206
Genomic Map