siRNA Details
| Gene Symbol: Cisd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025902.3 | NP_080178.1 | 3 | 1229 | ||
| GenBank mRNA | AK003486.1 | - | 3 | 1200 | ||
| AK005184.1 | - | 3 | 1188 | |||
| AM162549.1 | - | 3 | 1721 | |||
| BC025614.1 | - | 1 | 311 | |||
| BC058279.1 | - | 3 | 1229 | 6528467 | ||

Cisd2
Genomic Map