siRNA Details
| Gene Symbol: Ttc23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168475.1 | NP_001161947.1 | 2 | 618 | ||
| NM_001168477.1 | NP_001161949.1 | 2 | 618 | |||
| NM_025905.3 | NP_080181.2 | 2 | 618 | |||
| GenBank mRNA | AK141754.1 | - | 2 | 604 | ||
| AK165626.1 | - | 2 | 623 | |||
| AK166555.1 | - | 2 | 518 | |||
| AK166613.1 | - | 2 | 518 | |||
| AK211807.1 | - | 1 | 68 | |||
| BC038179.1 | - | 2 | 589 | 4485073 | ||

Ttc23
Genomic Map