siRNA Details
| Gene Symbol: Ttc23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168475.1 | NP_001161947.1 | 3, 4 | 768 | ||
| NM_001168476.1 | NP_001161948.1 | 2, 3 | 568 | |||
| NM_001168477.1 | NP_001161949.1 | 3, 4 | 768 | |||
| NM_025905.3 | NP_080181.2 | 3, 4 | 768 | |||
| GenBank mRNA | AK005431.1 | - | 2, 3 | 529 | ||
| AK076802.1 | - | 2, 3 | 362 | |||
| AK141754.1 | - | 3, 4 | 754 | |||
| AK165626.1 | - | 3, 4 | 773 | |||
| AK166555.1 | - | 3, 4 | 668 | |||
| AK166613.1 | - | 3, 4 | 668 | |||
| BC038179.1 | - | 3, 4 | 739 | 4485073 | ||

Ttc23
Genomic Map