siRNA Details
| Gene Symbol: Fam210b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025912.4 | NP_080188.3 | 3 | 955 | ||
| GenBank mRNA | AK008190.1 | - | 3 | 895 | ||
| AK076087.1 | - | 3 | 898 | |||
| AK084415.1 | - | 3 | 908 | |||
| AK147935.1 | - | 3 | 914 | |||
| AK148213.1 | - | 3 | 906 | |||
| AK148334.1 | - | 3 | 894 | |||
| AK148435.1 | - | 3 | 896 | |||
| AK149497.1 | - | 3 | 893 | |||
| BC016210.1 | - | 3 | 929 | 4924329 | ||
| BC057642.1 | - | 3 | 915 | 2655111 | ||

Fam210b
Genomic Map