siRNA Details
| Gene Symbol: Fam210b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025912.4 | NP_080188.3 | 3 | 965 | ||
| GenBank mRNA | AK008190.1 | - | 3 | 905 | ||
| AK076087.1 | - | 3 | 908 | |||
| AK084415.1 | - | 3 | 918 | |||
| AK147935.1 | - | 3 | 924 | |||
| AK148213.1 | - | 3 | 916 | |||
| AK148334.1 | - | 3 | 904 | |||
| AK148435.1 | - | 3 | 906 | |||
| AK149497.1 | - | 3 | 903 | |||
| BC016210.1 | - | 3 | 939 | 4924329 | ||
| BC057642.1 | - | 3 | 925 | 2655111 | ||

Fam210b
Genomic Map