siRNA Details
| Gene Symbol: Tmem246 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025944.3 | NP_080220.2 | 2 | 1079 | ||
| GenBank mRNA | AK013226.1 | - | 2 | 1008 | ||
| AK038514.1 | - | 2 | 1018 | |||
| AK039788.1 | - | 2 | 1013 | |||
| AK042679.1 | - | 2 | 1086 | |||
| AK045523.1 | - | 2 | 933 | |||
| AK075994.1 | - | 2 | 963 | |||
| AK165526.1 | - | 2 | 1018 | |||
| BC013800.1 | - | 2 | 924 | 2650271 | ||

Tmem246
Genomic Map