siRNA Details
| Gene Symbol: Polr3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164082.1 | NP_001157554.1 | 8 | 1822 | ||
| NM_025945.3 | NP_080221.3 | 9 | 1854 | |||
| GenBank mRNA | AK007614.1 | - | 1 | 127 | ||
| AK013165.1 | - | 8 | 1739 | |||
| AK156587.1 | - | 9 | 1806 | |||
| BC016102.1 | - | 9 | 1830 | 4911475 | ||

Polr3d
Genomic Map