siRNA Details
| Gene Symbol: Psmc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025959.3 | NP_080235.2 | 7 | 512 | ||
| GenBank mRNA | AK008996.1 | - | 256 | |||
| AK012174.1 | - | 7 | 513 | |||
| AK014354.1 | - | 7 | 500 | |||
| AK144728.1 | - | 509 | ||||
| AK166961.1 | - | 7 | 497 | |||
| BC025134.1 | - | 3 | 200 | |||
| BC037099.1 | - | 7 | 510 | |||
| BC043044.1 | - | 491 | ||||
| BC057997.1 | - | 468 | ||||
| BC116752.1 | - | 7 | 512 | 40126071 | ||
| BC119168.1 | - | 7 | 512 | 8733917 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001100509, FQ227742, FQ213949, FQ214043, BC107950, and others.

Psmc6
Genomic Map