siRNA Details
| Gene Symbol: Ccdc159 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164614.1 | NP_001158086.1 | 5, 6 | 760 | ||
| NM_025977.3 | NP_080253.2 | 5, 6 | 746 | |||
| GenBank mRNA | AK011114.1 | - | 5, 6 | 747 | ||
| AK016073.1 | - | 5, 6 | 571 | |||
| AK042884.1 | - | 5, 6 | 612 | |||
| BC120702.1 | - | 5, 6 | 721 | 40129625 | ||
| BC120704.1 | - | 5, 6 | 721 | 40129627 | ||
| BC145014.1 | - | 5, 6 | 737 | |||

Ccdc159
Genomic Map