siRNA Details
| Gene Symbol: Ndufa6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025987.3 | NP_080263.1 | 3 | 353 | ||
| GenBank mRNA | AK002749.1 | - | 3 | 300 | ||
| AK010988.1 | - | 3 | 302 | |||
| AK012328.1 | - | 3 | 302 | |||
| AV087995.1 | - | 2 | 90 | |||
| BC048451.1 | - | 3 | 353 | 6773780 | ||

Ndufa6
Genomic Map