siRNA Details
| Gene Symbol: Gp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025989.3 | NP_080265.2 | 3 | 332 | ||
| GenBank mRNA | AK009323.1 | - | 3 | 333 | ||
| AK009661.1 | - | 3 | 333 | |||
| AK009843.1 | - | 3 | 286 | |||
| BC119573.2 | - | 3 | 327 | 40057995 | ||

Gp2
Genomic Map