siRNA Details
| Gene Symbol: 2610301B20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026005.3 | NP_080281.3 | 5 | 553 | ||
| GenBank mRNA | AK011950.1 | - | 5 | 482 | ||
| AK146358.1 | - | 5 | 474 | |||
| BC058777.1 | - | 5 | 443 | 5397173 | ||
| BM948390.1 | - | 5 | 425 | |||
| CX220600.1 | - | 5 | 553 | |||

2610301B20Rik
Genomic Map