siRNA Details
| Gene Symbol: 2610301B20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026005.3 | NP_080281.3 | 6 | 1202 | ||
| GenBank mRNA | AK003887.1 | - | 1 | 617 | ||
| AK011950.1 | - | 6 | 1131 | |||
| AK146358.1 | - | 6 | 1123 | |||
| BC058777.1 | - | 6 | 1092 | 5397173 | ||

2610301B20Rik
Genomic Map