siRNA Details
| Gene Symbol: Mrps24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026080.2 | NP_080356.1 | 4 | 657 | ||
| GenBank mRNA | AK003117.1 | - | 4 | 641 | ||
| AK014099.1 | - | 4 | 644 | |||
| AK215321.1 | - | 1 | 284 | |||
| BC029262.1 | - | 4 | 631 | 4911699 | ||

Mrps24
Genomic Map