siRNA Details
| Gene Symbol: Gigyf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031408.2 | NP_113596.2 | 17, 18 | 2114 | ||
| GenBank mRNA | AK220289.1 | - | 10, 11 | 1123 | ||
| AY176042.1 | - | 17, 18 | 2130 | |||
| BC052875.1 | - | 11, 12 | 1218 | |||

Gigyf1
Genomic Map