siRNA Details
| Gene Symbol: Fancg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163233.1 | NP_001156705.1 | 7 | 1020 | ||
| NM_053081.2 | NP_444311.1 | 7 | 1020 | |||
| GenBank mRNA | AF112439.1 | - | 7 | 829 | ||
| AY049715.1 | - | 7 | 820 | |||
| BC050890.1 | - | 7 | 1277 | |||
| BC139763.1 | - | 7 | 823 | 40110334 | ||
| BC141282.1 | - | 7 | 836 | 9055894 | ||
| BE292126.1 | - | 2 | 124 | |||

Fancg
Genomic Map