siRNA Details
| Gene Symbol: Slc5a4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133184.2 | NP_573447.2 | 14 | 1792 | ||
| GenBank mRNA | AF251267.1 | - | 14 | 1792 | ||
| AF498304.1 | - | 3 | 363 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc5a4a| Gene Symbol: Slc5a4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133184.2 | NP_573447.2 | 14 | 1792 | ||
| GenBank mRNA | AF251267.1 | - | 14 | 1792 | ||
| AF498304.1 | - | 3 | 363 | |||