siRNA Details
Gene Symbol: Hiatl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083901.1 | NP_001077370.1 | 5, 6 | 665 | ||
NM_133680.3 | NP_598441.3 | 4, 5 | 762 | |||
GenBank mRNA | AK146596.1 | - | 5, 6 | 658 | ||
BC033469.1 | - | 4, 5 | 724 | 4951631 | ||
CN709077.1 | - | 1, 2 | 91 |
