siRNA Details
| Gene Symbol: Tspan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133681.4 | NP_598442.1 | 6 | 633 | ||
| GenBank mRNA | AK007428.1 | - | 6 | 631 | ||
| AK078869.1 | - | 6 | 634 | |||
| AK162521.1 | - | 6 | 633 | |||
| BC003448.1 | - | 6 | 573 | 2654254 | ||

Tspan1
Genomic Map