siRNA Details
| Gene Symbol: Fam13a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153574.2 | NP_705802.1 | 13 | 1574 | ||
| GenBank mRNA | AJ510136.1 | - | 13 | 1560 | ||
| AK036291.1 | - | 6 | 781 | |||
| AK052421.1 | - | 13 | 1515 | |||
| BC005554.1 | - | 13 | 1479 | |||
| BC054053.1 | - | 13 | 1548 | |||
| BC060978.1 | - | 13 | 1511 | |||
| BC080861.1 | - | 13 | 1574 | 30357292 | ||

Fam13a
Genomic Map