siRNA Details
| Gene Symbol: Sv2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109753.1 | NP_001103223.1 | 4 | 1339 | ||
| NM_153579.4 | NP_705807.2 | 4 | 1546 | |||
| GenBank mRNA | AK029736.1 | - | 5 | 1371 | ||
| AK043830.1 | - | 4 | 1266 | |||
| AK122359.1 | - | 3 | 491 | |||
| AK139478.1 | - | 4 | 1241 | |||
| AK140104.1 | - | 4 | 1259 | |||
| AK147261.1 | - | 4 | 1333 | |||
| AK159028.1 | - | 4 | 1244 | |||
| BC060224.1 | - | 3 | 930 | 30356990 | ||
| BC079568.1 | - | 4 | 1546 | |||

Sv2b
Genomic Map