siRNA Details
| Gene Symbol: Emc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175101.3 | NP_780310.1 | 8 | 1651 | ||
| GenBank mRNA | AK075595.1 | - | 8 | 1652 | ||
| AK167819.1 | - | 8 | 1500 | |||
| AK183927.1 | - | 1 | 118 | |||
| AK184001.1 | - | 1 | 118 | |||
| AK209735.1 | - | 1 | 65 | |||
| AK213208.1 | - | 1 | 118 | |||
| AK213644.1 | - | 1 | 35 | |||
| BC004641.1 | - | 8 | 1645 | 3157147 | ||

Emc3
Genomic Map