siRNA Details
| Gene Symbol: Fam53c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175104.4 | NP_780313.1 | 5 | 2365 | ||
| GenBank mRNA | AK044469.1 | - | 5 | 2169 | ||
| AK135712.1 | - | 4 | 2111 | |||
| AK154214.1 | - | 5 | 2169 | |||
| AK166841.1 | - | 4 | 2109 | |||
| BC044896.1 | - | 4 | 2090 | |||
| BC057111.1 | - | 5 | 2365 | 6417613 | ||

Fam53c
Genomic Map