siRNA Details
| Gene Symbol: Tmem177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175106.3 | NP_780315.1 | 2 | 1458 | ||
| GenBank mRNA | AK009043.1 | - | 2 | 1463 | ||
| AK076108.1 | - | 2 | 1464 | |||
| BC094344.1 | - | 2 | 1458 | 6402386 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem177| Gene Symbol: Tmem177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175106.3 | NP_780315.1 | 2 | 1458 | ||
| GenBank mRNA | AK009043.1 | - | 2 | 1463 | ||
| AK076108.1 | - | 2 | 1464 | |||
| BC094344.1 | - | 2 | 1458 | 6402386 | ||