siRNA Details
| Gene Symbol: 2310022A10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175107.5 | NP_780316.3 | 10 | 1692 | ||
| GenBank mRNA | AK163050.1 | - | 352 | |||
| AK210330.1 | - | 1 | 124 | |||
| BC023369.1 | - | 10 | 1671 | 5038597 | ||
| BX522780.1 | - | 1 | 194 | |||

2310022A10Rik
Genomic Map