siRNA Details
| Gene Symbol: Rps19bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175109.3 | NP_780318.1 | 4 | 741 | ||
| GenBank mRNA | AB233218.1 | - | 4 | 734 | ||
| AK075974.1 | - | 4 | 722 | |||
| BC038691.1 | - | 4 | 727 | 1495043 | ||
| BC050115.1 | - | 4 | 730 | 30017673 | ||
| BC065381.1 | - | 3 | 671 | |||

Rps19bp1
Genomic Map