siRNA Details
| Gene Symbol: Fbxl12os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK044040.1 | - | 2 | 1195 | ||
| BC147302.1 | - | 1 | 907 | 9056334 | ||
| BC147303.1 | - | 1 | 907 | 9056351 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxl12os| Gene Symbol: Fbxl12os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK044040.1 | - | 2 | 1195 | ||
| BC147302.1 | - | 1 | 907 | 9056334 | ||
| BC147303.1 | - | 1 | 907 | 9056351 | ||