siRNA Details
| Gene Symbol: Hspbap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175111.3 | NP_780320.2 | 3 | 364 | ||
| GenBank mRNA | AA879889.1 | - | 3 | 366 | ||
| AK017731.1 | - | 3 | 330 | |||
| AK028824.1 | - | 3 | 345 | |||
| AK049404.1 | - | 3 | 342 | |||
| AK076189.1 | - | 3 | 331 | |||
| AK089941.1 | - | 3 | 379 | |||
| AK138906.1 | - | 3 | 344 | |||

Hspbap1
Genomic Map