siRNA Details
| Gene Symbol: Trak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175114.3 | NP_780323.2 | 8, 9 | 1042 | ||
| GenBank mRNA | AK029123.1 | - | 9, 10 | 1440 | ||
| AK041436.1 | - | 9, 10 | 1410 | |||
| AK052264.1 | - | 9, 10 | 1429 | |||
| AK122426.1 | - | 9, 10 | 1132 | |||
| BC058971.1 | - | 8, 9 | 984 | 6832507 | ||
| BC116262.1 | - | 8, 9 | 1009 | |||
| BC116263.1 | - | 8, 9 | 1009 | |||
| BC129961.1 | - | 8, 9 | 1009 | |||
| BC129962.1 | - | 8, 9 | 1009 | |||

Trak1
Genomic Map