siRNA Details
| Gene Symbol: Snupn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178374.3 | NP_848461.1 | 3 | 313 | ||
| GenBank mRNA | AK002717.1 | - | 3 | 329 | ||
| AK013151.1 | - | 3 | 323 | |||
| AK155315.1 | - | 3 | 330 | |||
| BC023317.1 | - | 293 | ||||
| BC049638.1 | - | 3 | 313 | 6589249 | ||
| BC061258.1 | - | 3 | 318 | 30251661 | ||

Snupn
Genomic Map