siRNA Details
| Gene Symbol: Zfp869 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039965.1 | NP_001035054.1 | 5 | 780 | ||
| NM_001039967.1 | NP_001035056.1 | 6 | 888 | |||
| NM_181274.3 | NP_851791.1 | 5 | 848 | |||
| GenBank mRNA | AK004570.1 | - | 5 | 791 | ||
| AK030679.1 | - | 6 | 886 | |||
| AK054059.1 | - | 1 | 133 | |||
| AK083392.1 | - | 6 | 803 | |||
| AK088889.1 | - | 4 | 691 | |||
| AK136878.1 | - | 5 | 784 | |||
| BC063330.1 | - | 845 | ||||
| BC079868.1 | - | 7 | 1383 | |||

Zfp869
Genomic Map