siRNA Details
| Gene Symbol: Zfp869 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039965.1 | NP_001035054.1 | 5 | 1637 | ||
| NM_001039967.1 | NP_001035056.1 | 6 | 1745 | |||
| NM_181274.3 | NP_851791.1 | 5 | 1705 | |||
| GenBank mRNA | AK004570.1 | - | 5 | 1648 | ||
| AK030679.1 | - | 6 | 1743 | |||
| AK054059.1 | - | 1 | 990 | |||
| AK083392.1 | - | 6 | 1660 | |||
| AK088889.1 | - | 4 | 1548 | |||
| AK136878.1 | - | 5 | 1641 | |||
| BC063330.1 | - | 1702 | ||||
| BC079868.1 | - | 7 | 2240 | |||

Zfp869
Genomic Map