siRNA Details
| Gene Symbol: Wfdc21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183249.2 | NP_899072.1 | 1, 2 | 185 | ||
| GenBank mRNA | AA254806.1 | - | 1, 2 | 115 | ||
| AK003164.1 | - | 1, 2 | 127 | |||
| AK003352.1 | - | 1, 2 | 129 | |||
| BC096415.1 | - | 1, 2 | 120 | |||
| BY082582.1 | - | 1, 2 | 185 | |||
| BY094209.1 | - | 1, 2 | 127 | |||

Wfdc21
Genomic Map