siRNA Details
| Gene Symbol: Mga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164274.1 | NP_001157746.1 | 3 | 1943 | ||
| NM_013720.2 | NP_038748.2 | 3 | 1943 | |||
| GenBank mRNA | AF205935.1 | - | 2 | 1757 | ||
| AK032570.1 | - | 3 | 1924 | |||
| AK046252.1 | - | 3 | 1944 | |||
| AK132911.1 | - | 1 | 2220 | |||
| AK133775.1 | - | 3 | 2160 | |||
| AK139919.1 | - | 3 | 1895 | |||
| AK162302.1 | - | 3 | 1940 | |||
| AK172962.1 | - | 3 | 1866 | |||

Mga
Genomic Map