siRNA Details
| Gene Symbol: Slco1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013797.5 | NP_038825.1 | 14 | 2971 | ||
| GenBank mRNA | AF223067.1 | - | 14 | 2805 | ||
| AK041125.1 | - | 15 | 2909 | |||
| AY195868.1 | - | 14 | 2804 | |||
| AY195869.1 | - | 14 | 2804 | |||
| AY243579.1 | - | 14 | 2804 | |||
| BC041147.1 | - | 14 | 2753 | 4975986 | ||

Slco1a1
Genomic Map