siRNA Details
| Gene Symbol: Hspb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013868.4 | NP_038896.2 | 3 | 2017 | ||
| GenBank mRNA | AK030025.1 | - | 3 | 2018 | ||
| AK086011.1 | - | 3 | 1094 | |||
| BC046993.1 | - | 3 | 1053 | |||
| BC089584.1 | - | 3 | 1092 | 6755510 | ||

Hspb7
Genomic Map