siRNA Details
| Gene Symbol: RPP40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286132.1 | NP_001273061.1 | 4, 5 | 559 | ||
| NM_006638.3 | NP_006629.2 | 5, 6 | 628 | |||
| XM_011514253.2 | XP_011512555.1 | 5, 6 | 623 | |||
| GenBank mRNA | BC017871.1 | - | 4, 5 | 533 | 4278825 | |
| BM015573.1 | - | 5, 6 | 660 | |||
| BQ220155.1 | - | 5, 6 | 576 | |||

RPP40
Genomic Map