siRNA Details
| Gene Symbol: Rnf17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033043.1 | NP_001028215.1 | 3, 4 | 316 | ||
| GenBank mRNA | AF190166.1 | - | 3, 4 | 447 | ||
| AF285585.1 | - | 3, 4 | 316 | |||
| AK136196.1 | - | 3, 4 | 314 | |||
| AY854010.1 | - | 3, 4 | 316 | |||
| AY854011.1 | - | 3, 4 | 316 | |||

Rnf17
Genomic Map