siRNA Details
Gene Symbol: CPLX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006651.3 | NP_006642.1 | 4 | 1222 | ||
XM_011513391.1 | XP_011511693.1 | 3 | 1153 | |||
GenBank mRNA | AK126578.1 | - | 5 | 1094 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CPLX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006651.3 | NP_006642.1 | 4 | 1222 | ||
XM_011513391.1 | XP_011511693.1 | 3 | 1153 | |||
GenBank mRNA | AK126578.1 | - | 5 | 1094 |