siRNA Details
| Gene Symbol: Rnf19a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013923.2 | NP_038951.1 | 8 | 1815 | ||
| GenBank mRNA | AF120206.1 | - | 1708 | |||
| AF120207.1 | - | 7 | 1708 | |||
| AK147768.1 | - | 8 | 1803 | |||
| AK190444.1 | - | 2 | 55 | |||
| BC040769.1 | - | 7 | 1590 | 4457807 | ||
| DV656092.1 | - | 3 | 299 | |||
| X71642.1 | - | 4 | 362 | |||

Rnf19a
Genomic Map