siRNA Details
| Gene Symbol: Hspa14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015765.2 | NP_056580.2 | 2 | 241 | ||
| GenBank mRNA | AK031489.1 | - | 180 | |||
| AK031579.1 | - | 2 | 186 | |||
| AK088996.1 | - | 2 | 171 | |||
| AK145937.1 | - | 2 | 195 | |||
| AK150000.1 | - | 2 | 217 | |||
| AK153397.1 | - | 2 | 203 | |||
| BC002056.1 | - | 2 | 169 | 3493119 | ||
| BY147434.1 | - | 2 | 241 | |||
| U08215.1 | - | 2 | 201 | |||

Hspa14
Genomic Map