siRNA Details
| Gene Symbol: Sall2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015772.3 | NP_056587.2 | 1, 2 | 575 | ||
| GenBank mRNA | AB093233.1 | - | 1, 2 | 234 | ||
| AJ007396.1 | - | 1, 2 | 666 | |||
| BC054527.1 | - | 1, 2 | 270 | |||
| BC085361.1 | - | 1, 2 | 352 | 5706710 | ||
| CB525158.1 | - | 1, 2 | 357 | |||

Sall2
Genomic Map