siRNA Details
| Gene Symbol: Cfhr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015780.2 | NP_056595.1 | 4, 5 | 684 | ||
| GenBank mRNA | BC111870.1 | - | 4, 5 | 667 | 40057075 | |
| M29008.1 | - | 4, 5 | 684 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cfhr1| Gene Symbol: Cfhr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015780.2 | NP_056595.1 | 4, 5 | 684 | ||
| GenBank mRNA | BC111870.1 | - | 4, 5 | 667 | 40057075 | |
| M29008.1 | - | 4, 5 | 684 | |||