siRNA Details
Gene Symbol: Isg15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015783.3 | NP_056598.2 | 2 | 315 | ||
GenBank mRNA | BC031424.1 | - | 2 | 282 | 3670747 | |
BC083156.1 | - | 2 | 264 | 5338191 | ||
BC109346.2 | - | 2 | 265 | 40055493 | ||
CT010333.1 | - | 1 | 205 | |||
X56602.1 | - | 2 | 316 |
