siRNA Details
| Gene Symbol: Hist1h1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK002321.1 | - | 1003 | |||
| AK135614.1 | - | 736 | ||||
| AK155502.1 | - | 1005 | ||||
| BC089604.1 | - | 1004 | 6763721 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hist1h1c| Gene Symbol: Hist1h1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK002321.1 | - | 1003 | |||
| AK135614.1 | - | 736 | ||||
| AK155502.1 | - | 1005 | ||||
| BC089604.1 | - | 1004 | 6763721 | |||